Gene Transcripts

Gene Symbol
Preb
Entrez Gene ID
50907
Gene Name
prolactin regulatory element binding
Gene Aliases
C85705
Chromosome Location
Chr. 5: 30951667 - 30960386 on Build GRCm38
UniGene ID
Mm.272414
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001294302.1 NP_001281231.1 8 1861
NM_016703.3 NP_057912.2 9 2021
GenBank mRNA AF150808.1 - 9 1923
AF193017.1 - 9 1932
AK003323.1 - 1 596
AK013330.1 - 8 1815
AK029838.1 - 9 1957
AK036866.1 - 9 1959
AK151288.1 - 9 2003
BC017527.1 - 9 1924 4921119

View Full Product Details