Gene Transcripts
- Gene Symbol
- Atxn10
- Entrez Gene ID
- 54138
- Gene Name
- ataxin 10
- Gene Aliases
- AI325283, C77170, E46, Sca10, TEG-169, Tex169
- Chromosome Location
- Chr. 15: 85336381 - 85463836 on Build GRCm38
- UniGene ID
- Mm.248906
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_016843.3 |
NP_058539.2
|
7, 8 |
986 |
|
GenBank mRNA |
AK002504.1 |
- |
7, 8 |
1095 |
|
AK003530.1 |
- |
7, 8 |
913 |
|
AK047170.1 |
- |
7, 8 |
1036 |
|
AK049706.1 |
- |
7, 8 |
1030 |
|
AK077738.1 |
- |
7, 8 |
989 |
|
AK078459.1 |
- |
7, 8 |
1066 |
|
AK153271.1 |
- |
7, 8 |
988 |
|
AK161950.1 |
- |
|
1003 |
|
AK165941.1 |
- |
7, 8 |
1011 |
|
AK166899.1 |
- |
7, 8 |
1071 |
|
AK170507.1 |
- |
7, 8 |
1003 |
|
AK170751.1 |
- |
7, 8 |
1012 |
|
AK195052.1 |
- |
2, 3 |
220 |
|
AK217493.1 |
- |
|
48 |
|
BC002285.1 |
- |
1, 2 |
155 |
|
BC016410.1 |
- |
7, 8 |
996 |
3968016 |
BC046802.1 |
- |
7, 8 |
1122 |
6404607 |
X61506.1 |
- |
7, 8 |
1100 |
|
View Full Product Details