Gene Transcripts

Gene Symbol
Tmem9b
Entrez Gene ID
56786
Gene Name
TMEM9 domain family, member B
Gene Aliases
2310004K06Rik, AW539847, D7H11orf15, ICRFP703B1614Q5.3
Chromosome Location
Chr. 7: 109735834 - 109752870 on Build GRCm38
UniGene ID
Mm.240668
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020050.1 NP_064434.1 5 944
GenBank mRNA AK007430.1 - 5 912
AK009147.1 - 5 879
AK049882.1 - 5 1031
AK077253.1 - 5 900
AK147127.1 - 5 960
AK207353.1 - 1 275
AK215177.1 - 1 275
BC002208.1 - 5 928 3490055

View Full Product Details