Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_011706.2 | NP_035836.2 | 4 | 578 | |
GenBank mRNA | AB021665.1 | - | 4 | 660 | |
AK008689.1 | - | 2 | 103 | ||
AK089004.1 | - | 5 | 593 | ||
AK134052.1 | - | 3 | 891 | ||
AK162841.1 | - | 3 | 452 | ||
AK171222.1 | - | 3 | 501 | ||
AK200065.1 | - | 2 | 166 | ||
AK217648.1 | - | 2 | 77 | ||
BB631765.1 | - | 4 | 578 | ||
BC005415.1 | - | 3 | 525 | 3487527 |