Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_013782.4 | NP_038810.2 | 12 | 2023 | |
GenBank mRNA | AK036368.1 | - | 12 | 2024 | |
AK048026.1 | - | 12 | 2006 | ||
AK054435.1 | - | 1 | 220 | ||
AK081275.1 | - | 12 | 1990 | ||
AK137415.1 | - | 1984 | |||
BC009013.1 | - | 10 | 1690 | ||
BC053517.1 | - | 12 | 1960 | 6315175 |