Gene Transcripts
- Gene Symbol
- Bnip3l
- Entrez Gene ID
- 140923
- Gene Name
- BCL2/adenovirus E1B interacting protein 3-like
- Gene Aliases
- Nix
- Chromosome Location
- Chr. 15: 43643897 - 43667029 on Build Rnor_6.0
- UniGene ID
- Rn.827
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_080888.1 |
NP_543164.1
|
3, 4 |
361 |
|
XM_006252091.2 |
XP_006252153.1
|
3, 4 |
2162 |
|
XM_008770733.1 |
XP_008768955.1
|
3, 4 |
2291 |
|
XM_008770734.1 |
XP_008768956.1
|
3, 4 |
2292 |
|
GenBank mRNA |
AB077364.1 |
- |
3, 4 |
361 |
|
AF441118.1 |
- |
3, 4 |
430 |
|
BC081740.1 |
- |
3, 4 |
514 |
|
BC098658.1 |
- |
3, 4 |
395 |
7324593 |
FQ230363.1 |
- |
3, 4 |
354 |
|
FQ235252.1 |
- |
3, 4 |
380 |
|
View Full Product Details