Gene Transcripts

Gene Symbol
Bysl
Entrez Gene ID
359727
Gene Name
bystin-like
Gene Aliases
Chromosome Location
Chr. 9: 15393461 - 15403212 on Build Rnor_6.0
UniGene ID
Rn.185236
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_182674.3 NP_872615.3 3, 4 643
GenBank mRNA AY257675.4 - 3, 4 626
BC079030.1 - 3, 4 632 7114086

View Full Product Details