Gene Transcripts

Gene Symbol
BTN3A1
Entrez Gene ID
11119
Gene Name
butyrophilin subfamily 3 member A1
Gene Aliases
BT3.1, BTF5, BTN3.1, CD277
Chromosome Location
Chr. 6: 26402237 - 26415216 on Build GRCh38
UniGene ID
Hs.191510
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145008.1 NP_001138480.1 5 1045
NM_001145009.1 NP_001138481.1 5 1201
NM_007048.5 NP_008979.3 5 1201
NM_194441.2 NP_919423.1 5 1031
XM_005248833.2 XP_005248890.1 5 1169
XM_005248834.3 XP_005248891.1 5 1196
XM_006714983.2 XP_006715046.1 4 906
GenBank mRNA AK223474.1 - 4 982
AK290193.1 - 5 1198
AK291162.1 - 5 1031
AK295720.1 - 5 1188
AK298652.1 - 5 930
AK299327.1 - 5 987
BC118586.1 - 5 1162 40085016
BC121800.1 - 5 1162 40085015
BG744194.1 - 2 264
BM542639.1 - 574
U90552.2 - 5 1131
Y07827.1 - 5 923

View Full Product Details