Gene Transcripts

Gene Symbol
Apba2
Entrez Gene ID
83610
Gene Name
amyloid beta (A4) precursor protein-binding, family A, member 2
Gene Aliases
Mint2
Chromosome Location
Chr. 1: 125367273 - 125547741 on Build Rnor_6.0
UniGene ID
Rn.62687
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031780.1 NP_113968.1 4 1256
GenBank mRNA AF029107.1 - 4 1256

View Full Product Details