Gene Transcripts

Gene Symbol
Cabin1
Entrez Gene ID
94165
Gene Name
calcineurin binding protein 1
Gene Aliases
Cain
Chromosome Location
Chr. 20: 13836032 - 13963003 on Build Rnor_6.0
UniGene ID
Rn.4
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053575.2 NP_446027.2 9 1223
GenBank mRNA AF061947.1 - 9 1229

View Full Product Details