Gene Transcripts

Gene Symbol
Aagab
Entrez Gene ID
171435
Gene Name
alpha- and gamma-adaptin binding protein
Gene Aliases
P34
Chromosome Location
Chr. 8: 68526063 - 68564604 on Build Rnor_6.0
UniGene ID
Rn.163023
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_134398.2 NP_599225.1 8 771
XM_006243223.2 XP_006243285.1 8 800
GenBank mRNA AF178669.1 - 8 773
FQ224819.1 - 8 743

View Full Product Details