Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_012098.2 | NP_036230.1 | 2 | 1039 | |
XM_006717030.3 | XP_006717093.1 | 2 | 1077 | ||
GenBank mRNA | AF125175.1 | - | 1 | 560 | |
AK312876.1 | - | 2 | 958 | ||
AY358274.1 | - | 2 | 1058 | ||
BC012368.1 | - | 2 | 912 | 3865062 |