Gene Transcripts

Gene Symbol
Map6
Entrez Gene ID
29457
Gene Name
microtubule-associated protein 6
Gene Aliases
Mtap6
Chromosome Location
Chr. 1: 164225742 - 164292094 on Build Rnor_6.0
UniGene ID
Rn.37490
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017204.1 NP_058900.1 2, 3 1928
XM_006229755.2 XP_006229817.1 1, 2 2258
GenBank mRNA AJ002556.1 - 1, 2 1947
BC078848.1 - 1, 2 1806
X93495.1 - 2, 3 1928

View Full Product Details