Gene Transcripts

Gene Symbol
C1qb
Entrez Gene ID
29687
Gene Name
complement component 1, q subcomponent, B chain
Gene Aliases
Chromosome Location
Chr. 5: 155246444 - 155251995 on Build Rnor_6.0
UniGene ID
Rn.6702
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019262.1 NP_062135.1 3 399
GenBank mRNA FQ227589.1 - 3 324
FQ228182.1 - 3 259
X71127.1 - 3 399

View Full Product Details