Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_012328.2 | NP_036460.1 | 2 | 509 | |
GenBank mRNA | AB026908.1 | - | 2 | 342 | |
AF083247.1 | - | 2 | 343 | ||
AF093824.1 | - | 2 | 309 | ||
AK092204.1 | - | 3 | 367 | ||
AL080081.1 | - | 2 | 389 | ||
AY359045.1 | - | 1 | 163 | ||
BC028912.1 | - | 2 | 323 | 4943933 | |
CR533475.1 | - | 1 | 140 | ||
DA202575.1 | - | 2 | 509 |