Gene Transcripts

Gene Symbol
Rgs7
Entrez Gene ID
54296
Gene Name
regulator of G-protein signaling 7
Gene Aliases
Chromosome Location
Chr. 13: 93095659 - 93534452 on Build Rnor_6.0
UniGene ID
Rn.204219
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019343.1 NP_062216.1 2 173
GenBank mRNA AB024398.1 - 2 173

View Full Product Details