Gene Transcripts

Gene Symbol
SHANK2
Entrez Gene ID
22941
Gene Name
SH3 and multiple ankyrin repeat domains 2
Gene Aliases
AUTS17, CORTBP1, CTTNBP1, ProSAP1, SHANK, SPANK-3
Chromosome Location
Chr. 11: 70467856 - 71252724 on Build GRCh38
UniGene ID
Hs.268726
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012309.4 NP_036441.2 16, 17 2181
NM_133266.4 NP_573573.2 2, 3 378
NR_110766.1 2, 3 378
XM_005277932.3 XP_005277989.1 7, 8 1186
XM_017017387.1 XP_016872876.1 17, 18 2286
XM_017017388.1 XP_016872877.1 17, 18 2308
XM_017017389.1 XP_016872878.1 17, 18 2481
XM_017017390.1 XP_016872879.1 4, 5 3419
GenBank mRNA AB208025.1 - 16, 17 2147
AB208026.1 - 7, 8 969
AB208027.1 - 2, 3 356
AF141901.1 - 2, 3 270
BC093885.1 - 2, 3 378
BC112097.1 - 2, 3 378
BC113876.1 - 2, 3 301
BC114484.1 - 2, 3 300
BC143389.1 - 2, 3 378
DQ152234.1 - 15, 16 2069

View Full Product Details