Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001270645.1 | NP_001257574.1 | 5 | 821 | |
NM_001270647.1 | NP_001257576.1 | 5 | 1032 | ||
NM_017329.2 | NP_059025.2 | 5 | 818 | ||
GenBank mRNA | BC085353.1 | - | 5 | 821 | |
M15754.1 | - | 5 | 806 | ||
M33201.1 | - | 5 | 815 | ||
X13176.1 | - | 5 | 806 | ||
X13177.1 | - | 5 | 815 |