Gene Transcripts

Gene Symbol
PADI4
Entrez Gene ID
23569
Gene Name
peptidyl arginine deiminase 4
Gene Aliases
PAD, PAD4, PADI5, PDI4, PDI5
Chromosome Location
Chr. 1: 17308195 - 17364004 on Build GRCh38
UniGene ID
Hs.522969
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012387.2 NP_036519.2 6 609
XM_011541150.1 XP_011539452.1 4 421
XM_011541151.1 XP_011539453.1 6 609
XM_011541152.1 XP_011539454.1 3 141
XM_011541153.1 XP_011539455.1 6 609
XM_011541154.2 XP_011539456.1 6 609
XM_011541155.1 XP_011539457.1 6 609
XM_011541156.1 XP_011539458.1 6 609
XM_011541157.1 XP_011539459.1 4 267
GenBank mRNA AB017919.1 - 6 607
AK290507.1 - 6 607
AK309637.1 - 6 601
AK314839.1 - 6 609
BC025718.1 - 6 593 5228777

View Full Product Details