Gene Transcripts

Gene Symbol
Amy2a3
Entrez Gene ID
497039
Gene Name
amylase 2a3
Gene Aliases
Amy2
Chromosome Location
Chr. 2: 216373674 - 216382260 on Build Rnor_6.0
UniGene ID
Rn.67070
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031502.1 NP_113690.1 146
GenBank mRNA FQ233367.1 - 1 140
J00703.2 - 146

View Full Product Details