Gene Transcripts

Gene Symbol
FZD10
Entrez Gene ID
11211
Gene Name
frizzled class receptor 10
Gene Aliases
CD350, FZ-10, Fz10, FzE7, hFz10
Chromosome Location
Chr. 12: 130162459 - 130165740 on Build GRCh38
UniGene ID
Hs.31664
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007197.3 NP_009128.1 1 3055
GenBank mRNA AB027464.1 - 1 2589
BC070037.1 - 1 3052 30337870

View Full Product Details