Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001199054.1 | NP_001185983.1 | 5 | 645 | |
NM_001199055.1 | NP_001185984.1 | 5 | 496 | ||
NM_001199056.1 | NP_001185985.1 | 5 | 376 | ||
NM_013399.2 | NP_037531.2 | 5 | 613 | ||
GenBank mRNA | AF131218.1 | - | 4 | 692 | |
AF195661.1 | - | 5 | 608 | ||
AK292036.1 | - | 5 | 571 | ||
AK294257.1 | - | 5 | 466 | ||
AK302297.1 | - | 5 | 334 | ||
AK307067.1 | - | 6 | 574 | ||
AK311295.1 | - | 1 | 69 | ||
AL136698.1 | - | 5 | 571 | ||
AL833853.1 | - | 5 | 645 | ||
BC002882.2 | - | 5 | 559 | 3941342 | |
BC007604.1 | - | 1 | 147 | ||
CR533446.1 | - | 3 | 395 | ||
DQ167023.1 | - | 5 | 582 |