Gene Transcripts
- Gene Symbol
- C11orf58
- Entrez Gene ID
- 10944
- Gene Name
- chromosome 11 open reading frame 58
- Gene Aliases
- IMAGE145052, SMAP
- Chromosome Location
- Chr. 11: 16738601 - 16758354 on Build GRCh38
- UniGene ID
- Hs.368226
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_014267.5 |
NP_055082.1
|
2, 3 |
328 |
|
XM_017017143.1 |
XP_016872632.1
|
2, 3 |
2587 |
|
GenBank mRNA |
AK315382.1 |
- |
2, 3 |
282 |
|
BC007103.1 |
- |
2, 3 |
282 |
4281282 |
BC016352.1 |
- |
2, 3 |
286 |
4082845 |
BC020937.1 |
- |
2, 3 |
282 |
4281930 |
BC103747.1 |
- |
2, 3 |
309 |
6151644 |
BP210918.1 |
- |
3, 4 |
366 |
|
BP226882.1 |
- |
2, 3 |
528 |
|
BU661194.1 |
- |
3, 4 |
377 |
|
CR456882.1 |
- |
2, 3 |
150 |
|
U51678.1 |
- |
2, 3 |
185 |
|
View Full Product Details