Gene Transcripts

Gene Symbol
CARHSP1
Entrez Gene ID
23589
Gene Name
calcium regulated heat stable protein 1
Gene Aliases
CRHSP-24, CRHSP24, CSDC1
Chromosome Location
Chr. 16: 8852942 - 8869012 on Build GRCh38
UniGene ID
Hs.632184
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001042476.2 NP_001035941.1 4 686
NM_001278260.1 NP_001265189.1 5 494
NM_001278261.1 NP_001265190.1 5 633
NM_001278262.1 NP_001265191.1 5 592
NM_001278263.1 NP_001265192.1 4 368
NM_001278264.1 NP_001265193.1 4 510
NM_001278265.1 NP_001265194.1 4 502
NM_001278266.1 NP_001265195.1 4 478
NM_014316.3 NP_055131.2 4 380
XM_005255228.4 XP_005255285.3 4 544
XM_005255229.4 XP_005255286.1 4 356
XM_011522444.2 XP_011520746.1 4 355
GenBank mRNA AF115345.1 - 3 337
AK311777.1 - 4 374
BC001333.1 - 4 686
BC003366.1 - 4 377 2900952
BC108283.1 - 4 360 4430943
BM693675.1 - 4 510
BM920489.1 - 4 368
CA396216.1 - 4 437
DB101255.1 - 5 488
HY090855.1 - 4 380

View Full Product Details