Gene Transcripts

Gene Symbol
TRIM58
Entrez Gene ID
25893
Gene Name
tripartite motif containing 58
Gene Aliases
BIA2
Chromosome Location
Chr. 1: 247857199 - 247880138 on Build GRCh38
UniGene ID
Hs.269151
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015431.3 NP_056246.3 6 1723
GenBank mRNA AF327057.1 - 6 1723
AK096188.1 - 6 1605

View Full Product Details