Gene Transcripts

Gene Symbol
AGA
Entrez Gene ID
175
Gene Name
aspartylglucosaminidase
Gene Aliases
AGU, ASRG, GA
Chromosome Location
Chr. 4: 177430774 - 177442503 on Build GRCh38
UniGene ID
Hs.207776
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000027.3 NP_000018.2 9 1926
NM_001171988.1 NP_001165459.1 9 1896
NR_033655.1 8 1850

View Full Product Details