Gene Transcripts

Gene Symbol
ITLN1
Entrez Gene ID
55600
Gene Name
intelectin 1
Gene Aliases
HL-1, HL1, INTL, ITLN, LFR, hIntL, omentin
Chromosome Location
Chr. 1: 160876540 - 160885170 on Build GRCh38
UniGene ID
Hs.50813
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017625.2 NP_060095.2 8 932
GenBank mRNA AB036706.1 - 8 844
AF271386.1 - 8 855
AK000029.1 - 8 881
AK222533.1 - 8 919
AY065972.1 - 8 922
AY157361.1 - 8 934
AY157362.1 - 8 926
AY358359.1 - 8 918
AY549722.1 - 8 1021
AY619691.1 - 8 842
AY619692.1 - 8 899
BC020664.1 - 8 932 4693329
CR457224.1 - 7 816

View Full Product Details