Gene Transcripts

Gene Symbol
KIAA1549L
Entrez Gene ID
25758
Gene Name
KIAA1549 like
Gene Aliases
C11orf41, C11orf69, G2
Chromosome Location
Chr. 11: 33376569 - 33674102 on Build GRCh38
UniGene ID
Hs.502266
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_005252847.3 XP_005252904.2 2 443
XM_011519970.2 XP_011518272.1 2 443
XM_017017486.1 XP_016872975.1 2 443

View Full Product Details