Gene Transcripts

Gene Symbol
C6orf1
Entrez Gene ID
221491
Gene Name
chromosome 6 open reading frame 1
Gene Aliases
LBH
Chromosome Location
Chr. 6: 34246380 - 34249108 on Build GRCh38
UniGene ID
Hs.381300
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008703.2 NP_001008703.1 5 500
NM_001008704.2 NP_001008704.1 5 440
NM_001287396.1 NP_001274325.1 5 692
NM_001287397.1 NP_001274326.1 4 726
NM_001287398.1 NP_001274327.1 4 474
NM_178508.4 NP_848603.2 5 752
GenBank mRNA AK290164.1 - 5 434
BC023627.2 - 5 372 4764944
BC047919.1 - 5 633 6190218
BM474526.1 - 4 391
BU194178.1 - 5 573
BU956518.1 - 4 239
CK001844.1 - 5 503

View Full Product Details