Gene Transcripts

Gene Symbol
C7orf61
Entrez Gene ID
402573
Gene Name
chromosome 7 open reading frame 61
Gene Aliases
Chromosome Location
Chr. 7: 100456615 - 100464271 on Build GRCh38
UniGene ID
Hs.632306
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004323.1 NP_001004323.1 1 291
GenBank mRNA BC031966.1 - 1 291

View Full Product Details