Gene Transcripts

Gene Symbol
OR9K2
Entrez Gene ID
441639
Gene Name
olfactory receptor family 9 subfamily K member 2
Gene Aliases
OR12-2
Chromosome Location
Chr. 12: 55129769 - 55130776 on Build GRCh38
UniGene ID
Hs.553827
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005243.1 NP_001005243.1 1 884
GenBank mRNA BC136988.1 - 1 972

View Full Product Details