Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001282444.1 | NP_001269373.1 | 7 | 2126 | |
NM_001282445.1 | NP_001269374.1 | 6 | 1804 | ||
NM_006795.3 | NP_006786.2 | 5 | 1983 | ||
XM_011544739.1 | XP_011543041.1 | 6 | 1801 | ||
GenBank mRNA | AF001434.1 | - | 5 | 1934 | |
AF099011.1 | - | 1925 | |||
AK096747.1 | - | 1 | 837 | ||
AK097842.1 | - | 7 | 2091 | ||
AK098723.1 | - | 5 | 1188 | ||
AK124613.1 | - | 4 | 2901 | ||
AK126581.1 | - | 3 | 1561 | ||
AK297654.1 | - | 6 | 1662 | ||
AK299096.1 | - | 6 | 1715 | ||
AK300326.1 | - | 6 | 1764 | ||
AK302930.1 | - | 7 | 1405 | ||
BC104799.1 | - | 5 | 1875 | 8143802 | |
BC104825.1 | - | 5 | 1875 | 8143828 |