Gene Transcripts
- Gene Symbol
- C11orf49
- Entrez Gene ID
- 79096
- Gene Name
- chromosome 11 open reading frame 49
- Gene Aliases
- Chromosome Location
- Chr. 11: 46936524 - 47164381 on Build GRCh38
- UniGene ID
- Hs.368296
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001003676.2 |
NP_001003676.1
|
2, 3 |
259 |
|
NM_001003677.2 |
NP_001003677.1
|
2, 3 |
259 |
|
NM_001003678.2 |
NP_001003678.1
|
2, 3 |
259 |
|
NM_024113.4 |
NP_077018.1
|
2, 3 |
259 |
|
NR_103471.1 |
|
2, 3 |
259 |
|
XM_006718315.1 |
XP_006718378.1
|
2, 3 |
250 |
|
XM_011520365.2 |
XP_011518667.1
|
2, 3 |
250 |
|
XM_017018276.1 |
XP_016873765.1
|
2, 3 |
250 |
|
XM_017018278.1 |
XP_016873767.1
|
2, 3 |
251 |
|
XM_017018279.1 |
XP_016873768.1
|
2, 3 |
251 |
|
XM_017018280.1 |
XP_016873769.1
|
2, 3 |
187 |
|
XM_017018282.1 |
XP_016873771.1
|
2, 3 |
251 |
|
GenBank mRNA |
AK025863.1 |
- |
2, 3 |
168 |
|
AK315639.1 |
- |
2, 3 |
207 |
|
AK316318.1 |
- |
2, 3 |
251 |
|
AL136575.1 |
- |
2, 3 |
178 |
|
BC001860.2 |
- |
2, 3 |
205 |
3534541 |
BC009455.1 |
- |
2, 3 |
180 |
3535652 |
BC013988.2 |
- |
2, 3 |
170 |
3349493 |
BC052291.1 |
- |
2, 3 |
172 |
5936112 |
BQ428072.1 |
- |
3, 4 |
438 |
|
CR457351.1 |
- |
2, 3 |
142 |
|
DB027437.1 |
- |
2, 3 |
250 |
|
View Full Product Details