Gene Transcripts

Gene Symbol
OR4K1
Entrez Gene ID
79544
Gene Name
olfactory receptor family 4 subfamily K member 1
Gene Aliases
OR14-19
Chromosome Location
Chr. 14: 19920263 - 19936736 on Build GRCh38
UniGene ID
Hs.554584
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004063.2 NP_001004063.2 1 723
XM_011537153.2 XP_011535455.1 3 835
GenBank mRNA BC151145.1 - 1 723

View Full Product Details