Gene Transcripts

Gene Symbol
ZSWIM2
Entrez Gene ID
151112
Gene Name
zinc finger SWIM-type containing 2
Gene Aliases
MEX, ZZZ2
Chromosome Location
Chr. 2: 186827480 - 186849170 on Build GRCh38
UniGene ID
Hs.375054
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_182521.2 NP_872327.2 9 1243
GenBank mRNA AK128006.1 - 9 1243
BC031094.1 - 9 1242 5298288

View Full Product Details