Gene Transcripts

Gene Symbol
TSPAN10
Entrez Gene ID
83882
Gene Name
tetraspanin 10
Gene Aliases
OCSP
Chromosome Location
Chr. 17: 81637171 - 81648755 on Build GRCh38
UniGene ID
Hs.208219
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001290212.1 NP_001277141.1 3 785
NM_031945.4 NP_114151.3 2 551
NR_110866.1 2 551
GenBank mRNA AB593081.1 - 3 758
AB593082.1 - 2 546
AF325213.1 - 2 507
AK096926.1 - 2 533
BC021923.1 - 2 520 4876887
BC032802.1 - 2 522 5191718
BG760374.1 - 2 520
DA912496.1 - 2 533

View Full Product Details