Gene Transcripts

Gene Symbol
SHISA2
Entrez Gene ID
387914
Gene Name
shisa family member 2
Gene Aliases
C13orf13, PRO28631, TMEM46, WGAR9166, bA398O19.2, hShisa
Chromosome Location
Chr. 13: 26044597 - 26051060 on Build GRCh38
UniGene ID
Hs.433791
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001007538.1 NP_001007539.1 2 1814

View Full Product Details