Gene Transcripts

Gene Symbol
Asap3
Entrez Gene ID
230837
Gene Name
ArfGAP with SH3 domain, ankyrin repeat and PH domain 3
Gene Aliases
9430088F20Rik, Ddefl1, Gm140, UPLC1
Chromosome Location
Chr. 4: 136206349 - 136246578 on Build GRCm38
UniGene ID
Mm.77788
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008232.2 NP_001008233.1 14 1425
GenBank mRNA AK132294.1 - 14 1394
BC085250.1 - 14 1378 30730459

View Full Product Details