Gene Transcripts

Gene Symbol
C1qc
Entrez Gene ID
362634
Gene Name
complement component 1, q subcomponent, C chain
Gene Aliases
C1qg
Chromosome Location
Chr. 5: 155255013 - 155258631 on Build Rnor_6.0
UniGene ID
Rn.2393
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008524.1 NP_001008524.1 3 699
XM_006239225.1 XP_006239287.1 3 979
GenBank mRNA BC086409.1 - 3 699
FQ218827.1 - 3 723
FQ219610.1 - 3 751
FQ220269.1 - 3 751
FQ228978.1 - 3 983

View Full Product Details