Gene Transcripts

Gene Symbol
Ly6g6d
Entrez Gene ID
415062
Gene Name
lymphocyte antigen 6 complex, locus G6D
Gene Aliases
G6d, Ly6-D, Ng25
Chromosome Location
Chr. 20: 5061849 - 5064768 on Build Rnor_6.0
UniGene ID
Rn.145399
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001970.1 NP_001001970.1 2 116
XM_006256074.2 XP_006256136.1 2 452

View Full Product Details