Gene Transcripts

Gene Symbol
Cacybp
Entrez Gene ID
289144
Gene Name
calcyclin binding protein
Gene Aliases
Chromosome Location
Chr. 13: 77948774 - 77959089 on Build Rnor_6.0
UniGene ID
Rn.11540
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004208.1 NP_001004208.1 2 192
GenBank mRNA BC079007.1 - 2 192

View Full Product Details