Gene Transcripts

Gene Symbol
Saraf
Entrez Gene ID
290796
Gene Name
store-operated calcium entry-associated regulatory factor
Gene Aliases
Tmem66
Chromosome Location
Chr. 16: 61735189 - 61753438 on Build Rnor_6.0
UniGene ID
Rn.101865
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004213.1 NP_001004213.1 6 1533
GenBank mRNA BC078979.1 - 6 1533
FQ211074.1 - 1 376

View Full Product Details