Gene Transcripts

Gene Symbol
ZFP30
Entrez Gene ID
22835
Gene Name
ZFP30 zinc finger protein
Gene Aliases
ZNF745
Chromosome Location
Chr. 19: 37632488 - 37656261 on Build GRCh38
UniGene ID
Hs.716719
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001320666.1 NP_001307595.1 4, 5 758
NM_001320667.1 NP_001307596.1 4, 5 664
NM_001320668.1 NP_001307597.1 4, 5 534
NM_001320669.1 NP_001307598.1 4, 5 472
NM_014898.2 NP_055713.1 4, 5 705
GenBank mRNA AB023178.1 - 4, 5 705
BC041087.1 - 4, 5 734 5579247
BC050542.1 - 4, 5 663
GU388430.1 - 4, 5 534

View Full Product Details