Gene Transcripts

Gene Symbol
ATG14
Entrez Gene ID
22863
Gene Name
autophagy related 14
Gene Aliases
ATG14L, BARKOR, KIAA0831
Chromosome Location
Chr. 14: 55366391 - 55411858 on Build GRCh38
UniGene ID
Hs.414809
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014924.4 NP_055739.2 5 479
XM_011536563.2 XP_011534865.1 3 183
GenBank mRNA AK131251.1 - 5 479
AK292810.1 - 5 451
BC109089.1 - 5 464 40034573
BC109090.1 - 5 464 40034574

View Full Product Details