Gene Transcripts

Gene Symbol
Olr69
Entrez Gene ID
365326
Gene Name
olfactory receptor 69
Gene Aliases
Chromosome Location
Chr. 1: 168143384 - 168144322 on Build Rnor_6.0
UniGene ID
Rn.141914
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001280.1 NP_001001280.1 1 282

View Full Product Details