Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001168371.2 | NP_001161843.1 | 10 | 1578 | |
NM_001168372.2 | NP_001161844.1 | 10 | 1575 | ||
NM_001168373.2 | NP_001161845.1 | 10 | 1578 | ||
NM_027804.4 | NP_082080.3 | 10 | 1575 | ||
NM_145407.3 | NP_663382.2 | 9 | 1503 | ||
GenBank mRNA | AK043088.1 | - | 9 | 1339 | |
AK122396.2 | - | 9 | 1361 | ||
AK144099.1 | - | 5 | 605 | ||
AK146504.1 | - | 10 | 1401 | ||
AK148412.1 | - | 5 | 568 | ||
AK159473.1 | - | 10 | 1399 | ||
AK160807.1 | - | 10 | 1401 | ||
AK171942.1 | - | 10 | 1410 | ||
AK204964.1 | - | 1 | 21 | ||
BC006916.1 | - | 2 | 116 | ||
BC046824.1 | - | 1 | 38 | ||
BC060613.1 | - | 10 | 1503 | 6400986 |