Gene Transcripts

Gene Symbol
Btbd16
Entrez Gene ID
330660
Gene Name
BTB (POZ) domain containing 16
Gene Aliases
E330040A16Rik
Chromosome Location
Chr. 7: 130774069 - 130825899 on Build GRCm38
UniGene ID
Mm.271550
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081038.2 NP_001074507.2 3, 4 558
GenBank mRNA AK087912.1 - 3, 4 560
BB559682.2 - 1, 2 117
BB666983.1 - 3, 4 559

View Full Product Details