Gene Transcripts

Gene Symbol
ZNF423
Entrez Gene ID
23090
Gene Name
zinc finger protein 423
Gene Aliases
Ebfaz, JBTS19, NPHP14, OAZ, Roaz, ZFP423, Zfp104, hOAZ
Chromosome Location
Chr. 16: 49487531 - 49857919 on Build GRCh38
UniGene ID
Hs.443715
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271620.2 NP_001258549.1 4 1926
NM_015069.4 NP_055884.2 4 1816
XM_005255856.4 XP_005255913.1 4 2529
XM_005255857.4 XP_005255914.1 2 2019
XM_006721171.3 XP_006721234.1 7 2258
XM_017023076.1 XP_016878565.1 4 2543
XM_017023077.1 XP_016878566.1 4 1892
XM_017023078.1 XP_016878567.1 3 1865
GenBank mRNA AB018303.2 - 2 1583
AF221712.1 - 2 1583
AK309835.1 - 2 1601
AK314742.1 - 4 1927
BC112315.1 - 4 1818 8327783
BC112317.1 - 4 1818 8327785

View Full Product Details