Gene Transcripts

Gene Symbol
PLA2G3
Entrez Gene ID
50487
Gene Name
phospholipase A2 group III
Gene Aliases
GIII-SPLA2, SPLA2III, sPLA2-III
Chromosome Location
Chr. 22: 31134807 - 31140607 on Build GRCh38
UniGene ID
Hs.149623
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015715.4 NP_056530.2 4 1088
XM_011530204.1 XP_011528506.1 4 1310
XM_011530205.1 XP_011528507.1 4 1299
GenBank mRNA AF220490.1 - 4 1088
BC025316.1 - 4 964 5458919

View Full Product Details