Gene Transcripts

Gene Symbol
PID1
Entrez Gene ID
55022
Gene Name
phosphotyrosine interaction domain containing 1
Gene Aliases
HMFN2073, NYGGF4, P-CLI1, PCLI1
Chromosome Location
Chr. 2: 229023973 - 229271341 on Build GRCh38
UniGene ID
Hs.745038
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001100818.1 NP_001094288.1 3 618
NM_017933.4 NP_060403.3 4 829
XM_011511384.2 XP_011509686.1 3 501
XM_017004404.1 XP_016859893.1 4 5760
XM_017004405.1 XP_016859894.1 5 699
XM_017004406.1 XP_016859895.1 3 357
XM_017004407.1 XP_016859896.1 2 414
GenBank mRNA AB075874.1 - 4 484
AK000708.1 - 3 604
AK096636.1 - 3 560
AK125359.1 - 4 753
AY317148.1 - 3 450
BC040164.1 - 2 405 5589172

View Full Product Details